B-[3-(Tetrahydro-2H-pyran-4-yl)phenyl]boronic acid ≥97%

Supplier: AOBChem USA

Warning

310363-5G 310363-1G 310363-25G 310363-500MG
77632-628EA 1186 USD
77632-628 77637-966 77654-908 77645-942
B-[3-(Tetrahydro-2H-pyran-4-yl)phenyl]boronic acid ≥97%
B-[3-(Tetrahydro-2H-pyran-4-yl)phenyl]boronic acid

Precaución: For Research Use Only

Formula: C₁₁H₁₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD24448964
CAS Number: 2225175-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR