4'-Chloro-α-ethyl[1,1'-biphenyl]-4-methanol ≥95%

Supplier: AOBChem USA

Warning

310296-5G 310296-25G 310296-1G 310296-500MG
77628-526EA 1484.23 USD
77628-526 77635-626 77641-928 77645-940
4'-Chloro-α-ethyl[1,1'-biphenyl]-4-methanol ≥95%
4'-Chloro-α-ethyl[1,1'-biphenyl]-4-methanol

Precaución: For Research Use Only

Formula: 246.73 g/mol
MW: C₁₅H₁₅ClO
Storage Temperature: Ambient
MDL Number: MFCD35171873
CAS Number: 76431-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR