4'-Chloro-α,α-dimethyl[1,1'-biphenyl]-4-methanol ≥95%
Supplier: AOBChem USA
|
Warning
|
310290-5G
310290-25G
310290-1G
310290-500MG
77628-524EA
1484.23
USD
77628-524
77635-624
77641-926
77649-970
4'-Chloro-α,α-dimethyl[1,1'-biphenyl]-4-methanol ≥95%
4'-Chloro-α,α-dimethyl[1,1'-biphenyl]-4-methanol
Precaución: For Research Use Only
Formula:
246.73 g/mol MW: C₁₅H₁₅ClO Storage Temperature: Ambient |
MDL Number:
MFCD35171869 CAS Number: 92553-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...