1-(4'-Chloro[1,1'-biphenyl]-4-yl)-2-methyl-1-propanone ≥95%

Supplier: AOBChem USA

Warning

310274-500MG 310274-5G 310274-1G
77652-630EA 462 USD
77652-630 77654-902 77641-920
1-(4'-Chloro[1,1'-biphenyl]-4-yl)-2-methyl-1-propanone ≥95%
1-(4'-Chloro[1,1'-biphenyl]-4-yl)-2-methyl-1-propanone

Precaución: For Research Use Only

Formula: 258.74 g/mol
MW: C₁₆H₁₅ClO
Storage Temperature: Ambient
MDL Number: MFCD11924316
CAS Number: 58158-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR