4-Chloro-3-ethylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

31002-5G 31002-500MG 31002-25G 31002-1G
77632-624EA 674 USD
77632-624 77652-628 77654-900 77641-918
4-Chloro-3-ethylbenzyl alcohol ≥97%
4-Chloro-3-ethylbenzyl alcohol

Precaución: For Research Use Only

Formula: C₉H₁₁ClO
Storage Temperature: Ambient
MDL Number: MFCD11111049
CAS Number: 1314929-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR