4-Methoxy-α,α-dimethyl-3-(trifluoromethyl)benzenemethanol ≥95%
Supplier: AOBChem USA
|
Warning
|
309753-5G
309753-1G
309753-25G
309753-500MG
77632-620EA
1320
USD
77632-620
77637-962
77658-616
77645-930
4-Methoxy-α,α-dimethyl-3-(trifluoromethyl)benzenemethanol ≥95%
4-Methoxy-α,α-dimethyl-3-(trifluoromethyl)benzenemethanol
Precaución: For Research Use Only
Formula:
234.22 g/mol MW: C₁₁H₁₃F₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD35171707 CAS Number: 1783618-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...