1-[4-Methoxy-3-(trifluoromethyl)phenyl]-2-methyl-1-propanone ≥95%

Supplier: AOBChem USA

Warning

309749-5G 309749-1G 309749-500MG
77628-520EA 1484.23 USD
77628-520 77637-960 77645-924
1-[4-Methoxy-3-(trifluoromethyl)phenyl]-2-methyl-1-propanone ≥95%
1-[4-Methoxy-3-(trifluoromethyl)phenyl]-2-methyl-1-propanone

Precaución: For Research Use Only

Formula: 246.23 g/mol
MW: C₁₂H₁₃F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD28104498
CAS Number: 2173095-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR