Cyclopropyl[4-methoxy-3-(trifluoromethyl)phenyl]methanone ≥95%

Supplier: AOBChem USA

Warning

309748-5G 309748-1G 309748-500MG
77654-894EA 1983 USD
77654-894 77641-910 77649-968
Cyclopropyl[4-methoxy-3-(trifluoromethyl)phenyl]methanone ≥95%
Cyclopropyl[4-methoxy-3-(trifluoromethyl)phenyl]methanone

Precaución: For Research Use Only

Formula: 244.21 g/mol
MW: C₁₂H₁₁F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD28410790
CAS Number: 2173112-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR