4'-Ethoxy-3'-(trifluoromethyl)acetophenone ≥95%

Supplier: AOBChem USA

Warning

309727-5G 309727-25G 309727-1G 309727-500MG
77634-710EA 608 USD
77634-710 77654-892 77641-906 77645-920
4'-Ethoxy-3'-(trifluoromethyl)acetophenone ≥95%
4'-Ethoxy-3'-(trifluoromethyl)acetophenone

Precaución: For Research Use Only

Formula: C₁₁H₁₁F₃O₂
MW: 232.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13196590
CAS Number: 851263-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR