2-Bromo-6-fluoro-3-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

30955-5G 30955-25G 30955-1G 30955-500MG
77632-618EA 648 USD
77632-618 77654-890 77645-918 77649-966
2-Bromo-6-fluoro-3-(methylthio)benzaldehyde ≥97%
2-Bromo-6-fluoro-3-(methylthio)benzaldehyde

Precaución: For Research Use Only

Formula: 249.1 g/mol
MW: C₈H₆BrFOS
Storage Temperature: Ambient
MDL Number: MFCD29088237
CAS Number: 1879026-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR