3,4-Difluoro-2-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

30948-5G 30948-25G 30948-1G 30948-500MG
77632-616EA 796 USD
77632-616 77658-612 77641-904 77649-964
3,4-Difluoro-2-methylbenzyl alcohol ≥97%
3,4-Difluoro-2-methylbenzyl alcohol

Precaución: For Research Use Only

Formula: C₈H₈F₂O
MDL Number: MFCD07777158
CAS Number: 847502-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR