5-Bromo-4-chloro-2,3-difluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

30942-1G 30942-5G 30942-500MG
77637-958EA 619 USD
77637-958 77658-610 77645-916
5-Bromo-4-chloro-2,3-difluorophenylboronic acid ≥97%
5-Bromo-4-chloro-2,3-difluorophenylboronic acid

Precaución: For Research Use Only

Formula: C₆H₃BBrClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD29088228
CAS Number: 2121515-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR