6-Bromo-2,4-fifluoro-3-formylphenylboronic acid pinacol ester ≥95%
Supplier: AOBChem USA
|
Warning
|
30935-5G
30935-25G
30935-1G
30935-500MG
77634-708EA
299
USD
77634-708
77658-606
77641-902
77649-962
6-Bromo-2,4-fifluoro-3-formylphenylboronic acid pinacol ester ≥95%
6-Bromo-2,4-fifluoro-3-formylphenylboronic acid pinacol ester
Precaución: For Research Use Only
Formula:
346.96 g/mol MW: C₁₃H₁₄BBrF₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD29088226 CAS Number: 2121512-44-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...