6-Bromo-2,4-fifluoro-3-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

30934-5G 30934-25G
77634-706EA 160 USD
77634-706 77658-604
6-Bromo-2,4-fifluoro-3-formylphenylboronic acid ≥97%
6-Bromo-2,4-fifluoro-3-formylphenylboronic acid

Precaución: For Research Use Only

Formula: 264.817 g/mol
MW: C₇H₄BBrF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD29088225
CAS Number: 2121513-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR