S-benzyl benzothioate ≥95%

Supplier: AOBChem USA

Warning

309184-5G 309184-25G 309184-1G 309184-500MG
77628-514EA 1484.23 USD
77628-514 77635-622 77637-954 77649-960
S-benzyl benzothioate ≥95%
S-benzyl benzothioate

Precaución: For Research Use Only

Formula: C₁₄H₁₂OS
Storage Temperature: Ambient
MDL Number: MFCD00223101
CAS Number: 13402-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR