5-Bromo-6-chloropyridin-2-ol ≥95%

Supplier: AOBChem USA

Warning

30912-5G 30912-25G 30912-1G 30912-500MG
77634-704EA 585 USD
77634-704 77654-888 77641-900 77649-958
5-Bromo-6-chloropyridin-2-ol ≥95%
5-Bromo-6-chloropyridin-2-ol

Precaución: For Research Use Only

Formula: 208.44 g/mol
MW: C₅H₃BrClNO
Storage Temperature: Ambient
MDL Number: MFCD16606309
CAS Number: 1227603-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR