4-(Benzyloxycarbonyl)-3-methylphenylboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
30866-5G
30866-500MG
30866-25G
30866-1G
77632-614EA
933
USD
77632-614
77652-626
77658-602
77641-898
4-(Benzyloxycarbonyl)-3-methylphenylboronic acid pinacol ester ≥97%
4-(Benzyloxycarbonyl)-3-methylphenylboronic acid pinacol ester
Precaución: For Research Use Only
Formula:
352.232 g/mol MW: C₂₁H₂₅BO₄ Storage Temperature: Ambient |
MDL Number:
MFCD29088219 CAS Number: 2121511-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...