6-Methyl-1,3-benzodioxole-5-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

30864-5G 30864-1G 30864-25G
77634-702EA 195 USD
77634-702 77637-952 77658-600
6-Methyl-1,3-benzodioxole-5-carbaldehyde ≥95%
6-Methyl-1,3-benzodioxole-5-carbaldehyde

Precaución: For Research Use Only

Formula: C₉H₈O₃
MW: 164.158 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04041885
CAS Number: 58343-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR