6-Fluoro-5-methoxypicolinic acid ≥97%

Supplier: AOBChem USA

Warning

308485-1G 308485-5G 308485-500MG
77637-950EA 746 USD
77637-950 77654-886 77649-956
6-Fluoro-5-methoxypicolinic acid ≥97%
6-Fluoro-5-methoxypicolinic acid

Precaución: For Research Use Only

Formula: C₇H₆FNO₃
Storage Temperature: Ambient
MDL Number: MFCD18257410
CAS Number: 1211532-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR