3-Ethoxy-5-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

30829-5G 30829-25G 30829-1G 30829-500MG
77632-612EA 933 USD
77632-612 77654-884 77641-896 77649-954
3-Ethoxy-5-methoxybenzoic acid ≥97%
3-Ethoxy-5-methoxybenzoic acid

Precaución: For Research Use Only

Formula: 196.2 g/mol
MW: C₁₀H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD20643909
CAS Number: 62502-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR