5-Chloro-2,6-dimethoxypyridine-3-methanol ≥97%

Supplier: AOBChem USA

Warning

30825-5G 30825-1G 30825-25G 30825-500MG
77632-610EA 1406 USD
77632-610 77637-948 77654-882 77649-952
5-Chloro-2,6-dimethoxypyridine-3-methanol ≥97%
5-Chloro-2,6-dimethoxypyridine-3-methanol

Precaución: For Research Use Only

Formula: C₈H₁₀ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD29088217
CAS Number: 1879026-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR