4-Chloro-2,3-difluoro-5-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

30791-5G 30791-1G 30791-25G 30791-500MG
77632-608EA 1380 USD
77632-608 77637-946 77654-880 77649-950
4-Chloro-2,3-difluoro-5-hydroxybenzaldehyde ≥97%
4-Chloro-2,3-difluoro-5-hydroxybenzaldehyde

Precaución: For Research Use Only

Formula: 192.547 g/mol
MW: C₇H₃ClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD30529803
CAS Number: 2056110-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR