2-Bromo-1-fluoro-3-propoxybenzene ≥95%

Supplier: AOBChem USA

Warning

307908-5G 307908-500MG 307908-25G 307908-1G
77632-606EA 950 USD
77632-606 77652-624 77658-598 77641-894
2-Bromo-1-fluoro-3-propoxybenzene ≥95%
2-Bromo-1-fluoro-3-propoxybenzene

Precaución: For Research Use Only

Formula: C₉H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD30666686
CAS Number: 2158422-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR