1,3-Dibromo-2-fluoro-4-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

307905-5G 307905-1G 307905-500MG
77628-512EA 1484.23 USD
77628-512 77641-892 77645-912
1,3-Dibromo-2-fluoro-4-methoxybenzene ≥95%
1,3-Dibromo-2-fluoro-4-methoxybenzene

Precaución: For Research Use Only

Formula: C₇H₅Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD27957293
CAS Number: 1526600-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR