4-Chloro-2,3-difluoro-5-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30789-5G 30789-25G 30789-1G 30789-500MG
77632-604EA 1289 USD
77632-604 77654-878 77641-890 77645-910
4-Chloro-2,3-difluoro-5-methoxyphenylboronic acid pinacol ester ≥97%
4-Chloro-2,3-difluoro-5-methoxyphenylboronic acid pinacol ester

Precaución: For Research Use Only

Formula: 304.525 g/mol
MW: C₁₃H₁₆BClF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD30529802
CAS Number: 2121511-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR