3-Ethoxy-5-methoxyphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

30774-5G 30774-1G 30774-25G 30774-500MG
77632-602EA 1193 USD
77632-602 77637-944 77654-876 77649-948
3-Ethoxy-5-methoxyphenylboronic acid ≥95%
3-Ethoxy-5-methoxyphenylboronic acid

Precaución: For Research Use Only

Formula: C₉H₁₃BO₄
Storage Temperature: Ambient
MDL Number: MFCD29088204
CAS Number: 2121511-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR