5-Bromo-2,6-dimethoxypyridine-3-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

30740-5G 30740-1G 30740-25G 30740-500MG
77632-598EA 749 USD
77632-598 77637-940 77658-594 77649-946
5-Bromo-2,6-dimethoxypyridine-3-carboxylic acid ≥95%
5-Bromo-2,6-dimethoxypyridine-3-carboxylic acid

Precaución: For Research Use Only

Formula: 262.057 g/mol
MW: C₈H₈BrNO₄
Storage Temperature: Ambient
MDL Number: MFCD29088188
CAS Number: 1826110-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR