5-Bromo-2,6-dimethoxypyridine-3-boronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
30734-5G
30734-25G
30734-1G
30734-500MG
77634-700EA
321
USD
77634-700
77654-872
77645-902
77645-904
5-Bromo-2,6-dimethoxypyridine-3-boronic acid ≥97%
5-Bromo-2,6-dimethoxypyridine-3-boronic acid
Precaución: For Research Use Only
Formula:
261.866 g/mol MW: C₇H₉BBrNO₄ Storage Temperature: Ambient |
MDL Number:
MFCD29088183 CAS Number: 2121513-61-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...