1-(3-Fluoro-2,5-dimethylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

306690-1G 306690-5G 306690-500MG
77637-938EA 591 USD
77637-938 77654-870 77645-898
1-(3-Fluoro-2,5-dimethylphenyl)ethanone ≥95%
1-(3-Fluoro-2,5-dimethylphenyl)ethanone

Precaución: For Research Use Only

Formula: 166.192 g/mol
MW: C₁₀H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD28552606
CAS Number: 1087765-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR