1-Bromo-4-chloro-5-iodo-2-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

306502-5G 306502-25G 306502-1G 306502-500MG
77632-594EA 1357 USD
77632-594 77658-590 77641-884 77645-896
1-Bromo-4-chloro-5-iodo-2-methoxybenzene ≥95%
1-Bromo-4-chloro-5-iodo-2-methoxybenzene

Precaución: For Research Use Only

Formula: 347.375 g/mol
MW: C₇H₅BrClIO
Storage Temperature: Ambient
MDL Number: MFCD34704354
CAS Number: 2386247-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR