Isopropyl 2-isopropoxynicotinate ≥95%

Supplier: AOBChem USA

Warning

30597-5G 30597-25G 30597-1G 30597-500MG
77632-588EA 933 USD
77632-588 77658-588 77641-882 77645-890
Isopropyl 2-isopropoxynicotinate ≥95%
Isopropyl 2-isopropoxynicotinate

Precaución: For Research Use Only

Formula: C₁₂H₁₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD23113743
CAS Number: 1395038-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR