Methyl 2-(propan-2-yloxy)pyridine-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

30596-5G 30596-25G 30596-1G 30596-500MG
77632-586EA 933 USD
77632-586 77654-862 77641-880 77649-942
Methyl 2-(propan-2-yloxy)pyridine-3-carboxylate ≥97%
Methyl 2-(propan-2-yloxy)pyridine-3-carboxylate

Precaución: For Research Use Only

Formula: 195.215 g/mol
MW: C₁₀H₁₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD23115032
CAS Number: 1030829-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR