2-Chloro-5-fluoro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

30557-5G 30557-1G 30557-25G 30557-500MG
77634-698EA 492 USD
77634-698 77637-932 77654-860 77649-940
2-Chloro-5-fluoro-3-methylbenzaldehyde ≥97%
2-Chloro-5-fluoro-3-methylbenzaldehyde

Precaución: For Research Use Only

Formula: C₈H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD28751752
CAS Number: 1807026-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR