Methyl 2,4,6-trifluoro-α-oxobenzeneacetate ≥97%

Supplier: AOBChem USA

Warning

305567-5G 305567-1G 305567-500MG
77628-506EA 1484.23 USD
77628-506 77641-878 77649-938
Methyl 2,4,6-trifluoro-α-oxobenzeneacetate ≥97%
Methyl 2,4,6-trifluoro-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: C₉H₅F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD26057459
CAS Number: 1564586-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR