Methyl 2,4,6-trichloro-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

305566-5G 305566-25G 305566-1G 305566-500MG
77628-504EA 1484.23 USD
77628-504 77635-620 77637-930 77645-888
Methyl 2,4,6-trichloro-α-oxobenzeneacetate ≥95%
Methyl 2,4,6-trichloro-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: 267.49 g/mol
MW: C₉H₅Cl₃O₃
Storage Temperature: Ambient
MDL Number: MFCD29930775
CAS Number: 1599538-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR