Methyl 5-chloro-2-fluoro-3-methyl-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

305564-5G 305564-1G 305564-500MG
77654-858EA 2053 USD
77654-858 77641-876 77645-886
Methyl 5-chloro-2-fluoro-3-methyl-α-oxobenzeneacetate ≥95%
Methyl 5-chloro-2-fluoro-3-methyl-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: 230.62 g/mol
MW: C₁₀H₈ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD34892402
CAS Number: 2090978-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR