Methyl 2,5-dichloro-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

305516-5G 305516-1G 305516-500MG
77628-502EA 1484.23 USD
77628-502 77641-874 77645-882
Methyl 2,5-dichloro-α-oxobenzeneacetate ≥95%
Methyl 2,5-dichloro-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: 233.05 g/mol
MW: C₉H₆Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD26057462
CAS Number: 1565067-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR