Methyl 5-chloro-2-fluoro-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

305511-1G 305511-5G 305511-500MG
77637-926EA 591 USD
77637-926 77658-586 77649-936
Methyl 5-chloro-2-fluoro-α-oxobenzeneacetate ≥95%
Methyl 5-chloro-2-fluoro-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: 216.59 g/mol
MW: C₉H₆ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD34892372
CAS Number: 1890250-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR