Methyl 2,5-difluoro-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

305500-5G 305500-1G 305500-25G 305500-500MG
77632-584EA 989 USD
77632-584 77637-924 77654-854 77649-934
Methyl 2,5-difluoro-α-oxobenzeneacetate ≥95%
Methyl 2,5-difluoro-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: C₉H₆F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD19315221
CAS Number: 1267231-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR