Methyl 5-bromo-2-fluoro-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

305491-5G 305491-1G 305491-500MG
77628-500EA 1484.23 USD
77628-500 77641-872 77645-880
Methyl 5-bromo-2-fluoro-α-oxobenzeneacetate ≥95%
Methyl 5-bromo-2-fluoro-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: 261.04 g/mol
MW: C₉H₆BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD28107724
CAS Number: 1352420-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR