Methyl 2,3-difluoro-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

305404-5G 305404-1G 305404-500MG
77628-498EA 1484.23 USD
77628-498 77637-920 77652-618
Methyl 2,3-difluoro-α-oxobenzeneacetate ≥95%
Methyl 2,3-difluoro-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: 200.14 g/mol
MW: C₉H₆F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD28105866
CAS Number: 1192312-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR