Methyl 2,3-dichloro-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

305389-5G 305389-1G 305389-500MG
77628-496EA 1484.23 USD
77628-496 77637-918 77645-878
Methyl 2,3-dichloro-α-oxobenzeneacetate ≥95%
Methyl 2,3-dichloro-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: 233.05 g/mol
MW: C₉H₆Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD28104885
CAS Number: 1646328-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR