Methyl 2-fluoro-5-methyl-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

305388-5G 305388-1G 305388-500MG
77628-494EA 1484.23 USD
77628-494 77641-870 77645-876
Methyl 2-fluoro-5-methyl-α-oxobenzeneacetate ≥95%
Methyl 2-fluoro-5-methyl-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: C₁₀H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD34892310
CAS Number: 2155652-70-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR