Methyl 2,6-difluoro-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

305380-5G 305380-1G 305380-500MG
77628-492EA 1484.23 USD
77628-492 77637-916 77649-930
Methyl 2,6-difluoro-α-oxobenzeneacetate ≥95%
Methyl 2,6-difluoro-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: 200.14 g/mol
MW: C₉H₆F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD28105934
CAS Number: 132116-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR