Methyl 8-bromo-2,3-dihydrobenzo[b][1,4]dioxine-5-carboxylate ≥95%

Supplier: AOBChem USA

Warning

304289-1G 304289-5G 304289-500MG
77634-694EA 805 USD
77634-694 77654-850 77649-926
Methyl 8-bromo-2,3-dihydrobenzo[b][1,4]dioxine-5-carboxylate ≥95%
Methyl 8-bromo-2,3-dihydrobenzo[b][1,4]dioxine-5-carboxylate

Precaución: For Research Use Only

Formula: C₁₀H₉BrO₄
Storage Temperature: Ambient
MDL Number: MFCD29919264
CAS Number: 823225-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR