2-Bromo-5-chloropyridine-3-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30421-5G 30421-25G 30421-1G 30421-500MG
77632-580EA 841 USD
77632-580 77654-848 77641-864 77645-872
2-Bromo-5-chloropyridine-3-boronic acid pinacol ester ≥97%
2-Bromo-5-chloropyridine-3-boronic acid pinacol ester

Precaución: For Research Use Only

Formula: C₁₁H₁₄BBrClNO₂
Storage Temperature: Ambient
MDL Number: MFCD18910598
CAS Number: 2121511-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR