1-Bromo-4-fluoro-2-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

303788-5G 303788-1G 303788-500MG
77628-488EA 1484.23 USD
77628-488 77637-914 77645-868
1-Bromo-4-fluoro-2-(trifluoromethoxy)benzene ≥97%
1-Bromo-4-fluoro-2-(trifluoromethoxy)benzene

Precaución: For Research Use Only

Formula: C₇H₃BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD25458129
CAS Number: 1242252-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR