4-Chloro-2,3-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

30359-5G 30359-1G 30359-25G 30359-500MG
77632-576EA 1198 USD
77632-576 77637-912 77658-580 77649-922
4-Chloro-2,3-dimethylbenzoic acid ≥97%
4-Chloro-2,3-dimethylbenzoic acid

Precaución: For Research Use Only

Formula: 184.62 g/mol
MW: C₉H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD18917145
CAS Number: 5628-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR