4-Chloro-2,3-dimethylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

30355-5G 30355-1G 30355-500MG
77658-578EA 1877 USD
77658-578 77641-860 77645-866
4-Chloro-2,3-dimethylphenylboronic acid ≥97%
4-Chloro-2,3-dimethylphenylboronic acid

Precaución: For Research Use Only

Formula: 184.428 g/mol
MW: C₈H₁₀BClO₂
Storage Temperature: Ambient
MDL Number: MFCD29088154
CAS Number: 2121511-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR