(3,5-Difluorophenyl)trimethylstannane ≥95%

Supplier: AOBChem USA

Warning

303213-5G 303213-1G 303213-500MG
77628-484EA 1484.23 USD
77628-484 77637-906 77645-862
(3,5-Difluorophenyl)trimethylstannane ≥95%
(3,5-Difluorophenyl)trimethylstannane

Precaución: For Research Use Only

Formula: 276.9 g/mol
MW: C₉H₁₂F₂Sn
Storage Temperature: Ambient
MDL Number: MFCD28402448
CAS Number: 129436-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR